Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008409.2 | NP_032435.2 | 3 | 494 | ||
GenBank mRNA | AK014024.1 | - | 3 | 262 | ||
AK051956.1 | - | 3 | 473 | |||
AK135229.1 | - | 3 | 446 | |||
AK167351.1 | - | 3 | 495 | |||
BC031420.1 | - | 3 | 461 | 3668557 | ||
BQ568915.1 | - | 3 | 462 | |||
L38971.1 | - | 3 | 497 |