Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008410.2 | NP_032436.1 | 4 | 627 | ||
GenBank mRNA | AB030203.1 | - | 4 | 647 | ||
AB030204.1 | - | 3 | 443 | |||
AK002599.1 | - | 4 | 590 | |||
AK003224.1 | - | 4 | 594 | |||
AK003587.1 | - | 4 | 616 | |||
AK003928.1 | - | 4 | 615 | |||
AK005125.1 | - | 4 | 618 | |||
AK076139.1 | - | 4 | 636 | |||
AK077482.1 | - | 4 | 629 | |||
AK088270.1 | - | 4 | 614 | |||
AK150378.1 | - | 4 | 647 | |||
AK150396.1 | - | 4 | 646 | |||
AK151714.1 | - | 4 | 646 | |||
AK151889.1 | - | 4 | 647 | |||
AK152068.1 | - | 4 | 635 | |||
AK152516.1 | - | 4 | 647 | |||
AK152650.1 | - | 4 | 647 | |||
AK152731.1 | - | 4 | 618 | |||
AK152983.1 | - | 4 | 639 | |||
AK153103.1 | - | 4 | 590 | |||
AK159307.1 | - | 4 | 590 | |||
AK159340.1 | - | 4 | 615 | |||
AK159378.1 | - | 4 | 646 | |||
AK159426.1 | - | 4 | 615 | |||
AK159619.1 | - | 4 | 639 | |||
AK159628.1 | - | 4 | 615 | |||
AK159792.1 | - | 4 | 615 | |||
AK159962.1 | - | 4 | 618 | |||
AK160101.1 | - | 4 | 615 | |||
AK160725.1 | - | 4 | 646 | |||
AK194069.1 | - | 192 | ||||
BC004731.1 | - | 4 | 598 | 3583154 | ||
BC010320.1 | - | 4 | 630 | 3599190 | ||
BC021786.1 | - | 4 | 636 | 4458918 | ||
U76253.1 | - | 4 | 627 |