Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008410.2 | NP_032436.1 | 5 | 825 | ||
GenBank mRNA | AB030203.1 | - | 5 | 845 | ||
AB030204.1 | - | 4 | 641 | |||
AK002599.1 | - | 5 | 788 | |||
AK003224.1 | - | 5 | 792 | |||
AK003587.1 | - | 5 | 814 | |||
AK003928.1 | - | 5 | 812 | |||
AK005125.1 | - | 5 | 816 | |||
AK076139.1 | - | 5 | 834 | |||
AK077482.1 | - | 5 | 827 | |||
AK150378.1 | - | 5 | 845 | |||
AK150396.1 | - | 5 | 844 | |||
AK151714.1 | - | 5 | 844 | |||
AK151889.1 | - | 5 | 845 | |||
AK152068.1 | - | 5 | 833 | |||
AK152516.1 | - | 5 | 845 | |||
AK152650.1 | - | 5 | 845 | |||
AK152731.1 | - | 5 | 816 | |||
AK152983.1 | - | 5 | 837 | |||
AK153103.1 | - | 5 | 788 | |||
AK159307.1 | - | 5 | 788 | |||
AK159340.1 | - | 5 | 813 | |||
AK159426.1 | - | 5 | 813 | |||
AK159619.1 | - | 5 | 837 | |||
AK159628.1 | - | 5 | 813 | |||
AK159792.1 | - | 5 | 813 | |||
AK159962.1 | - | 5 | 816 | |||
AK160101.1 | - | 5 | 813 | |||
AK160725.1 | - | 5 | 844 | |||
AK200695.1 | - | 1 | 48 | |||
AK210559.1 | - | 1 | 48 | |||
BC004731.1 | - | 5 | 796 | 3583154 | ||
BC010320.1 | - | 5 | 828 | 3599190 | ||
BC021786.1 | - | 5 | 834 | 4458918 | ||
U76253.1 | - | 5 | 825 |