Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ivl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008412.3 | NP_032438.2 | 2 | 172 | ||
GenBank mRNA | AA789977.1 | - | 2 | 152 | ||
BC152326.1 | - | 2 | 152 | 40141142 |