Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004984.2 | NP_004975.2 | 3 | 487 | ||
GenBank mRNA | AB210045.1 | - | 3 | 312 | ||
AB290174.1 | - | 3 | 553 | |||
AK315202.1 | - | 3 | 507 | |||
AL707807.1 | - | 3 | 521 | |||
BC140009.1 | - | 3 | 540 | |||
BC146670.1 | - | 3 | 490 | 40148192 | ||
BC150208.1 | - | 3 | 490 | 40148890 | ||
U06698.1 | - | 3 | 427 |