Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008444.4 | NP_032470.3 | 9 | 3105 | ||
GenBank mRNA | AK083700.1 | - | 9 | 3093 | ||
AK122258.1 | - | 9 | 3066 | |||
AK147467.1 | - | 9 | 3099 | |||
AK148200.1 | - | 9 | 3086 | |||
AK161946.1 | - | 7 | 1544 | |||
AK171262.1 | - | 1 | 492 | |||
BC006767.1 | - | 1 | 388 | |||
BC065132.1 | - | 3 | 1417 | |||
BC139388.1 | - | 8 | 3024 | 8862410 | ||
BC139389.1 | - | 8 | 3024 | 8862411 | ||
D26077.1 | - | 8 | 3018 |