Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008446.2 | NP_032472.2 | 2, 3 | 647 | ||
GenBank mRNA | AK084837.1 | - | 2, 3 | 624 | ||
AK135179.1 | - | 2, 3 | 595 | |||
AK135430.1 | - | 2, 3 | 141 | |||
AK171938.1 | - | 2, 3 | 561 | |||
BC050946.1 | - | 2, 3 | 585 | 6822283 | ||
BC054537.1 | - | 2, 3 | 569 | |||
D12646.1 | - | 2, 3 | 974 |