Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008447.4 | NP_032473.2 | 28 | 4155 | ||
GenBank mRNA | BC058396.1 | - | 28 | 3872 | 6824963 |