Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004988.4 | NP_004979.3 | 3 | 543 | ||
GenBank mRNA | AK314979.1 | - | 3 | 543 | ||
AY148486.1 | - | 3 | 501 | |||
BC017555.1 | - | 3 | 510 | 3922845 | ||
BP314195.1 | - | 2 | 464 | |||
BT009789.1 | - | 1 | 323 | |||
DB053667.1 | - | 3 | 557 |