Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170851.1 | NP_001164322.1 | 4 | 426 | ||
NM_008462.5 | NP_032488.4 | 4 | 426 | |||
GenBank mRNA | AF253057.1 | - | 3 | 260 | ||
AF253058.1 | - | 3 | 260 | |||
AF253059.1 | - | 3 | 260 | |||
AF395446.1 | - | 3 | 260 | |||
AK017140.1 | - | 4 | 417 | |||
AY860971.1 | - | 3 | 260 | |||
BC064711.1 | - | 4 | 426 | 30295755 | ||
EF025059.1 | - | 3 | 260 | |||
U10304.1 | - | 4 | 354 |