Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008465.5 | NP_032491.2 | 13 | 1482 | ||
GenBank mRNA | AK028259.1 | - | 13 | 1483 | ||
AK028307.1 | - | 13 | 1501 | |||
AK154800.1 | - | 13 | 1500 | |||
AK169311.1 | - | 13 | 1505 | |||
AK169348.1 | - | 13 | 1505 | |||
AK203891.1 | - | 2 | 149 | |||
BC006771.1 | - | 13 | 1467 | 3588534 | ||
U20619.1 | - | 13 | 1487 | |||
U34228.1 | - | 12 | 1331 |