Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008473.2 | NP_032499.2 | 2 | 814 | ||
GenBank mRNA | AK019521.1 | - | 2 | 815 | ||
BC117842.1 | - | 2 | 781 | 40093577 | ||
BC117843.1 | - | 2 | 781 | 40093580 | ||
M10937.1 | - | 2 | 822 |