Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY028607.1 | - | 4, 5 | 951 | ||
BC114971.1 | - | 4, 5 | 947 | 40039508 |