Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002257.1 | - | 8 | 1242 | ||
AK019391.1 | - | 8 | 1279 | |||
AK131637.1 | - | 8 | 1267 | |||
BC046755.1 | - | 8 | 1264 | 5693226 | ||
BC087888.1 | - | 5 | 661 | |||
X51905.1 | - | 8 | 1213 |