Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008498.2 | NP_032524.1 | 3 | 1773 | ||
GenBank mRNA | AK141086.1 | - | 1775 | |||
BC092374.1 | - | 3 | 1906 | 30363447 | ||
S68107.1 | - | 6 | 1305 | |||
Z27410.1 | - | 3 | 1660 |