Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008522.3 | NP_032548.2 | 5 | 600 | ||
GenBank mRNA | AK036491.1 | - | 5 | 601 | ||
AK144556.1 | - | 5 | 601 | |||
AK150799.1 | - | 5 | 601 | |||
AK151822.1 | - | 5 | 601 | |||
AK164781.1 | - | 5 | 602 | |||
BC006904.1 | - | 5 | 608 | 3600033 | ||
BC008530.1 | - | 5 | 581 | |||
CT010339.1 | - | 5 | 566 | |||
D88510.1 | - | 5 | 600 | |||
FJ538998.1 | - | 5 | 566 | |||
J03298.1 | - | 5 | 569 |