Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008533.2 | NP_032559.2 | 3 | 446 | ||
GenBank mRNA | AK089255.1 | - | 3 | 437 | ||
AK156548.1 | - | 3 | 437 | |||
D37797.1 | - | 3 | 416 |