Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amacr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008537.4 | NP_032563.2 | 5 | 1190 | ||
GenBank mRNA | AK002401.1 | - | 5 | 1158 | ||
AK048249.1 | - | 5 | 1191 | |||
AK160586.1 | - | 5 | 1193 | |||
BC015825.1 | - | 5 | 1118 | 3980103 | ||
U89906.1 | - | 5 | 1098 |