Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marcks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008538.2 | NP_032564.1 | 2 | 2534 | ||
GenBank mRNA | AK136452.1 | - | 1 | 703 | ||
AK151646.1 | - | 1 | 970 | |||
AK153217.1 | - | 1 | 967 | |||
BC046601.1 | - | 2 | 2496 | 6311439 | ||
M60474.1 | - | 2 | 2524 |