Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Marcks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008538.2 | NP_032564.1 | 2 | 2045 | ||
GenBank mRNA | AK082623.1 | - | 1 | 196 | ||
AK136452.1 | - | 1 | 214 | |||
AK151646.1 | - | 1 | 481 | |||
AK153217.1 | - | 1 | 478 | |||
BC046601.1 | - | 2 | 2007 | 6311439 | ||
M60474.1 | - | 2 | 2035 |