Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Masp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008555.2 | NP_032581.2 | 2 | 138 | ||
GenBank mRNA | AB049755.1 | - | 2 | 151 | ||
AK031598.1 | - | 2 | 199 | |||
AK032944.1 | - | 2 | 145 | |||
AK043055.1 | - | 2 | 198 | |||
BC131637.1 | - | 2 | 138 | 40109637 | ||
BC131638.1 | - | 2 | 138 | 40109639 | ||
D16492.1 | - | 151 |