Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008562.3 | NP_032588.1 | 3 | 1971 | ||
GenBank mRNA | AK010424.1 | - | 2 | 1146 | ||
AK144229.1 | - | 1 | 521 | |||
AK148607.1 | - | 1 | 511 | |||
AK159504.1 | - | 3 | 1883 | |||
BC005427.1 | - | 3 | 1893 | 3493739 | ||
BC021638.1 | - | 3 | 1910 | 4190996 | ||
U35623.1 | - | 3 | 1883 |