Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008564.2 | NP_032590.2 | 5 | 816 | ||
GenBank mRNA | AF004105.1 | - | 4 | 706 | ||
AK037751.1 | - | 5 | 741 | |||
AK077801.1 | - | 5 | 720 | |||
AK088156.1 | - | 5 | 729 | |||
AK129039.1 | - | 5 | 697 | |||
AK145176.1 | - | 5 | 727 | |||
AK146187.1 | - | 5 | 731 | |||
AK146375.1 | - | 5 | 740 | |||
BC055318.1 | - | 5 | 725 | 5696348 | ||
D86725.1 | - | 5 | 736 |