Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008564.2 | NP_032590.2 | 8 | 1382 | ||
GenBank mRNA | AF004105.1 | - | 7 | 1272 | ||
AK037751.1 | - | 8 | 1307 | |||
AK077801.1 | - | 8 | 1287 | |||
AK088156.1 | - | 8 | 1295 | |||
AK088378.1 | - | 8 | 1282 | |||
AK129039.1 | - | 8 | 1263 | |||
AK145176.1 | - | 8 | 1294 | |||
AK146375.1 | - | 8 | 1306 | |||
BC055318.1 | - | 8 | 1291 | 5696348 | ||
D86725.1 | - | 8 | 1302 |