Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033196.1 | - | 9 | 1186 | ||
AK051795.1 | - | 9 | 1156 | |||
AK088558.1 | - | 2 | 102 | |||
AK133826.1 | - | 9 | 1187 | |||
AK209639.1 | - | 1 | 42 | |||
AK212108.1 | - | 1 | 42 | |||
AK214383.1 | - | 1 | 42 | |||
BC090645.1 | - | 9 | 1139 | |||
BC094416.1 | - | 9 | 1151 | 5707724 | ||
D26090.1 | - | 8 | 1206 |