Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008568.2 | NP_032594.1 | 15 | 2238 | ||
GenBank mRNA | AK076059.1 | - | 11 | 1688 | ||
AK077766.1 | - | 12 | 1797 | |||
AK132748.1 | - | 2802 | ||||
AK133091.1 | - | 15 | 2234 | |||
AK144980.1 | - | 15 | 2250 | |||
AK149913.1 | - | 15 | 2232 | |||
AK150059.1 | - | 15 | 2235 | |||
AK150066.1 | - | 15 | 2232 | |||
AK154052.1 | - | 15 | 2235 | |||
AK199009.1 | - | 1 | 116 | |||
AK199740.1 | - | 1 | 116 | |||
BC023232.1 | - | 15 | 2214 | |||
BC065164.1 | - | 15 | 2222 | 6829737 | ||
BC066024.1 | - | 15 | 2177 | 6829337 | ||
D26091.1 | - | 15 | 2238 |