Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008571.1 | NP_032597.1 | 2 | 161 | ||
GenBank mRNA | AK133521.1 | - | 2 | 162 | ||
AY007570.1 | - | 2 | 161 | |||
BC119248.1 | - | 2 | 154 | 8733997 | ||
BC119250.1 | - | 2 | 154 | 8733999 | ||
J05177.1 | - | 2 | 143 |