Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008574.3 | NP_032600.2 | 2 | 587 | ||
GenBank mRNA | AK077172.1 | - | 2 | 558 | ||
BC055104.1 | - | 2 | 557 | 6773089 | ||
BC061167.1 | - | 2 | 537 | 6771333 | ||
BY456161.1 | - | 1 | 130 | |||
M29603.1 | - | 2 | 560 |