Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mesp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008588.2 | NP_032614.2 | 2 | 825 | ||
GenBank mRNA | AK006394.1 | - | 2 | 256 | ||
BC012689.1 | - | 2 | 1034 | |||
BC125505.1 | - | 2 | 825 | 40130020 | ||
BC125507.1 | - | 2 | 825 | 40130022 | ||
CF105615.1 | - | 2 | 300 | |||
D83674.1 | - | 2 | 888 |