Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008592.2 | NP_032618.2 | 1 | 2543 | ||
GenBank mRNA | AF045017.1 | - | 1 | 2530 | ||
BC052011.1 | - | 1 | 2381 | 5720391 |