Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfge8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045489.1 | NP_001038954.1 | 4 | 583 | ||
NM_008594.2 | NP_032620.2 | 5 | 694 | |||
GenBank mRNA | AB021130.2 | - | 5 | 601 | ||
AB025280.1 | - | 4 | 490 | |||
AK087555.1 | - | 4 | 573 | |||
AK089211.1 | - | 4 | 549 | |||
AK145183.1 | - | 4 | 547 | |||
AK148371.1 | - | 4 | 572 | |||
AK152088.1 | - | 5 | 683 | |||
AK155047.1 | - | 5 | 692 | |||
AK169997.1 | - | 4 | 547 | |||
AK170935.1 | - | 4 | 547 | |||
AK171143.1 | - | 5 | 658 | |||
BC003892.1 | - | 4 | 527 | 3593310 | ||
BC003904.1 | - | 4 | 522 | 3601565 | ||
BC018577.1 | - | 4 | 520 | |||
BC037475.1 | - | 1 | 909 | |||
M38337.1 | - | 5 | 646 | |||
Y11684.2 | - | 4 | 490 |