Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008598.2 | NP_032624.1 | 4 | 441 | ||
GenBank mRNA | AB092489.1 | - | 3 | 352 | ||
AB092490.1 | - | 3 | 352 | |||
AK090389.1 | - | 431 | ||||
BC031888.1 | - | 4 | 410 | 5372377 | ||
BC096557.1 | - | 4 | 444 | 1366095 | ||
M84524.1 | - | 3 | 367 |