Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008599.4 | NP_032625.2 | 4 | 525 | ||
GenBank mRNA | AK042017.1 | - | 4 | 527 | ||
AK042480.1 | - | 4 | 443 | |||
AK156012.1 | - | 4 | 526 | |||
AK156029.1 | - | 4 | 526 | |||
AK156453.1 | - | 4 | 526 | |||
AK157109.1 | - | 4 | 527 | |||
AK157118.1 | - | 4 | 526 | |||
BC003343.1 | - | 4 | 513 | 3257716 | ||
M34815.1 | - | 4 | 563 |