Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008600.4 | NP_032626.2 | 4 | 815 | ||
GenBank mRNA | AK053490.1 | - | 4 | 816 | ||
AK053494.1 | - | 4 | 778 | |||
AK136287.1 | - | 4 | 816 | |||
BC082567.1 | - | 4 | 804 | 30551292 | ||
U27502.1 | - | 4 | 771 |