Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008607.2 | NP_032633.1 | 10 | 2204 | ||
GenBank mRNA | AK150728.1 | - | 10 | 2191 | ||
AK151626.1 | - | 10 | 2191 | |||
AK152507.1 | - | 10 | 2190 | |||
AK152916.1 | - | 10 | 2191 | |||
AK159921.1 | - | 10 | 2191 | |||
AK160819.1 | - | 10 | 2189 | |||
X66473.1 | - | 10 | 2173 |