Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008608.3 | NP_032634.3 | 9 | 1610 | ||
GenBank mRNA | AK040968.1 | - | 1583 | |||
AK088476.1 | - | 9 | 1610 | |||
AK134340.1 | - | 9 | 1542 | |||
AK138611.1 | - | 9 | 1551 | |||
AK149636.1 | - | 9 | 1539 | |||
AK149907.1 | - | 9 | 1590 | |||
AK149988.1 | - | 9 | 1510 | |||
AK163601.1 | - | 9 | 1602 | |||
AK165014.1 | - | 9 | 1611 | |||
AK185474.1 | - | 1 | 40 | |||
AK186174.1 | - | 1 | 40 | |||
AK189497.1 | - | 2 | 158 | |||
AK194864.1 | - | 1 | 40 | |||
AK208886.1 | - | 1 | 40 | |||
AK212820.1 | - | 1 | 40 | |||
AK219528.1 | - | 1 | 40 | |||
BC076638.1 | - | 9 | 1602 | 30658833 | ||
DQ249870.1 | - | 9 | 1348 | |||
U54984.1 | - | 9 | 1594 | |||
X83536.2 | - | 9 | 1520 |