Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Me1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198933.1 | NP_001185862.1 | 3 | 513 | ||
NM_008615.2 | NP_032641.2 | 3 | 435 | |||
GenBank mRNA | AK004980.1 | - | 3 | 301 | ||
AK077968.1 | - | 3 | 303 | |||
AK163278.1 | - | 3 | 513 | |||
AK163403.1 | - | 3 | 513 | |||
BC003287.1 | - | 3 | 292 | 3582779 | ||
BC011081.1 | - | 3 | 333 | 3978265 | ||
BC080660.1 | - | 3 | 291 | 4240732 | ||
BC094028.1 | - | 3 | 290 | 4974808 | ||
CJ070715.1 | - | 3 | 436 | |||
J02652.1 | - | 3 | 336 | |||
M26756.1 | - | 3 | 278 |