Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ627041.1 | - | 1 | 86 | ||
AK036415.1 | - | 1 | 241 | |||
AK048532.1 | - | 1 | 271 | |||
AK081829.1 | - | 1 | 203 | |||
AK152379.1 | - | 1 | 185 | |||
AK165261.1 | - | 1 | 204 | |||
AK168352.1 | - | 1 | 152 | |||
BC013444.1 | - | 1 | 254 | 4221660 | ||
BC138314.1 | - | 1 | 144 | 8861334 | ||
BC138315.1 | - | 1 | 144 | 8861335 | ||
U38196.1 | - | 1 | 260 |