Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008624.3 | NP_032650.1 | 6 | 1358 | ||
GenBank mRNA | AB004879.1 | - | 6 | 1342 | ||
AF043581.1 | - | 5 | 955 | |||
AK050469.1 | - | 6 | 1202 | |||
AK164066.1 | - | 6 | 1002 | |||
BC024389.1 | - | 6 | 1075 | 4973582 |