Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004882.3 | NP_004873.3 | 5 | 369 | ||
GenBank mRNA | AF098297.1 | - | 5 | 276 | ||
AK292389.1 | - | 5 | 352 | |||
BC015040.1 | - | 5 | 323 | |||
BC021175.1 | - | 5 | 337 | |||
BC031018.1 | - | 5 | 300 | |||
BC038987.1 | - | 5 | 344 | |||
BC046098.2 | - | 5 | 320 | 5495320 | ||
BU596189.1 | - | 5 | 372 | |||
U03644.1 | - | 5 | 308 |