Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004888.3 | NP_004879.1 | 2 | 256 | ||
GenBank mRNA | AF038954.1 | - | 2 | 194 | ||
BC008452.1 | - | 2 | 182 | 4246202 | ||
BF692207.1 | - | 259 | ||||
BI596632.1 | - | 2 | 234 | |||
CR456971.1 | - | 2 | 131 | |||
CR542237.1 | - | 2 | 131 |