Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Laptm4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008640.2 | NP_032666.2 | 7 | 1611 | ||
GenBank mRNA | AK013963.1 | - | 6 | 629 | ||
AK075699.1 | - | 7 | 846 | |||
AK084515.1 | - | 7 | 1254 | |||
AK088958.1 | - | 7 | 871 | |||
AK165638.1 | - | 7 | 880 | |||
AK182919.1 | - | 4 | 393 | |||
BC086667.1 | - | 7 | 791 | 30633914 | ||
BC089021.1 | - | 7 | 824 | 6734038 | ||
U34259.1 | - | 8 | 1478 |