Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008657.2 | NP_032683.1 | 3 | 1066 | ||
GenBank mRNA | AK029370.1 | - | 3 | 1082 | ||
X59060.1 | - | 3 | 1066 |