Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252220.1 | NP_001239149.1 | 8 | 901 | ||
NM_001252222.1 | NP_001239151.1 | 8 | 724 | |||
NM_001252223.1 | NP_001239152.1 | 8 | 724 | |||
NM_008676.3 | NP_032702.1 | 8 | 724 | |||
GenBank mRNA | AF227188.1 | - | 8 | 853 | ||
AK128992.1 | - | 8 | 516 | |||
AK129043.1 | - | 8 | 494 | |||
AK154081.1 | - | 8 | 510 | |||
BC052709.1 | - | 8 | 514 | 6417055 | ||
BC129867.1 | - | 7 | 376 | 40110124 | ||
BC129868.1 | - | 7 | 376 | 40110128 | ||
CV558786.1 | - | 8 | 724 | |||
U73039.1 | - | 8 | 520 |