Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfe2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130450.1 | NP_001123922.1 | 2 | 1044 | ||
NM_008686.3 | NP_032712.2 | 2 | 1102 | |||
GenBank mRNA | AF071084.1 | - | 1 | 448 | ||
AK131183.1 | - | 2 | 1037 | |||
AK152040.1 | - | 2 | 1083 | |||
AK153529.1 | - | 2 | 1084 | |||
AK154167.1 | - | 2 | 1013 | |||
AK184212.1 | - | 1 | 112 | |||
AK201732.1 | - | 1 | 102 | |||
BC043063.1 | - | 1 | 684 | 5705669 | ||
BC047283.1 | - | 1 | 444 | 6489220 | ||
BC099895.1 | - | 2 | 1068 | |||
X78709.1 | - | 1 | 480 |