Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048168.2 | NP_001041633.1 | 10 | 1556 | ||
NM_001277095.1 | NP_001264024.1 | 9 | 1515 | |||
NM_008692.4 | NP_032718.2 | 10 | 1629 | |||
GenBank mRNA | BC020117.1 | - | 10 | 1555 | 4009737 | |
BC053723.1 | - | 10 | 1559 | 30053020 | ||
BC085261.1 | - | 10 | 1555 | 6397760 | ||
BE647422.1 | - | 1 | 131 | |||
BU559204.1 | - | 1 | 403 | |||
DV655537.1 | - | 3 | 602 |