Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008694.2 | NP_032720.2 | 4 | 778 | ||
GenBank mRNA | AK009649.1 | - | 4 | 732 | ||
AK165210.1 | - | 4 | 731 | |||
L37297.1 | - | 4 | 783 | |||
U95002.1 | - | 4 | 737 |