Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008695.2 | NP_032721.2 | 5 | 1702 | ||
GenBank mRNA | AB017202.1 | - | 5 | 1664 | ||
AK045306.1 | - | 5 | 1677 | |||
AK051664.1 | - | 5 | 1678 | |||
AK052839.1 | - | 5 | 1676 | |||
AK083629.1 | - | 5 | 1679 | |||
AK128978.1 | - | 2 | 824 | |||
AK140832.1 | - | 5 | 1676 | |||
AK164258.1 | - | 5 | 1673 | |||
AY074820.1 | - | 5 | 1663 | |||
BC054746.1 | - | 5 | 1684 | 6405073 | ||
BC057016.1 | - | 5 | 1630 | 5719452 |