Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmbr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008703.2 | NP_032729.1 | 3 | 1075 | ||
GenBank mRNA | BC119237.1 | - | 3 | 1144 | 8733986 | |
BC144884.1 | - | 4 | 1229 | |||
BC144885.1 | - | 2 | 796 |
Set Membership: |
G-protein coupled receptor |