Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nqo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008706.5 | NP_032732.3 | 4 | 511 | ||
GenBank mRNA | AK075568.1 | - | 4 | 512 | ||
BC004579.1 | - | 4 | 489 | 3586888 | ||
CT010284.1 | - | 3 | 378 | |||
S75951.1 | - | 4 | 585 | |||
U12961.1 | - | 4 | 486 |